Learn More
Electron Microscopy Sciences Applied Physics LaB6 Cathode for Leica Lithography SMVM
SDP
Catalog No. 50949845
Encompass
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten. Cambridge SMVM base.
Type: LaB6
Angle: 60°
Tips: 40 mm
Catalog No. 50-949-845 Supplier Electron Microscopy Sciences Supplier No. 80929L20S
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten. Cambridge SMVM base.
Type: LaB6
Angle: 60°
Tips: 40 mm