Learn More
Electron Microscopy Sciences Applied Physics LaB6 Cathode Leica Lithography MVM
SDP
Catalog No. 50949846
Encompass
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten. Cambridge MVM base.
Type: LaB6
Angle: 90°
Tips: 20 mm
Type: non-shunted
Catalog No. 50-949-846 Supplier Electron Microscopy Sciences Supplier No. 80929LCM
Lanthanum hexaboride (LaB6) and cerium hexaboride (CeB6) cathodes are ideal for many small spot size applications such as SEM, TEM, surface analysis and metrology, and for high current applications such as microwave tubes, lithography, electron-beam welders, X-ray sources and free electron lasers. Greater brightness from low work function yielding higher currents at lower temps than tungsten. Cambridge MVM base.
Type: LaB6
Angle: 90°
Tips: 20 mm
Type: non-shunted