Learn More
Electron Microscopy Sciences Leica Leica Microtome Blades, L/P, 75.74 x 8.09 x 0.27 mm
SDP
Catalog No. 5026902
Encompass_Preferred
Made from high quality stainless steel. High and low profile disposable blades for use with all Leica microtomes as well as the Cryocut and Cryostat. A teflon-fluorocarbon resin-coated edge allows sections to slide off the edge without distortion. They minimize compression. Comes 50 blades in a dispenser box. Low profile blade measures: 2.98"(L) x 0.318"(W) x 0.010”(T) (75.74 x 8.09 x 0.27 mm). High profile blade measures: 2.98”(L) x 0.557”(W) x 0.012”(T) (75.74 x 14.14 x 0.32 mm)
Dimensions: 75.74 x 8.09 x 0.27 mm
Catalog No. 50-269-02 Supplier Electron Microscopy Sciences Supplier No. 63065LP
Add to Cart
Add to Cart
Made from high quality stainless steel. High and low profile disposable blades for use with all Leica microtomes as well as the Cryocut and Cryostat. A teflon-fluorocarbon resin-coated edge allows sections to slide off the edge without distortion. They minimize compression. Comes 50 blades in a dispenser box. Low profile blade measures: 2.98"(L) x 0.318"(W) x 0.010”(T) (75.74 x 8.09 x 0.27 mm). High profile blade measures: 2.98”(L) x 0.557”(W) x 0.012”(T) (75.74 x 14.14 x 0.32 mm)
Dimensions: 75.74 x 8.09 x 0.27 mm