Learn More
Leica Biosystems Leica Paraplast Plus (8-1kg)
Supplier:  Leica Biosystems 39602004/DUP
Encompass_Preferred
Paraplast Plus is suggested for large tissue or tissue that may normally present infiltration issues. Paraplast Plus contains a small percentage of dimethyl sulfoxide (DMSO), allowing accelerated tissue penetration.
Catalog No. 50-255-2389