Learn More
Leica Biosystems Leica reticle for 13613532 eyepiece, 100 divisions
Catalog No. NC3833233
Encompass
Reticle designed to install in a compatible eyepiece to provide a 100-division measurement scale for transmitted-light microscopy. Intended for use with Leica 13613532 eyepiece (10x/20 focusing eyepiece), it enables precise linear measurement, particle counting, and consistent reference marking.
  • Provides 100 divisions for precise linear measurement and particle counting.
  • Compatible with focusing eyepieces that accept removable reticles.
  • Improves reproducibility and reference marking during routine microscopy.
  • Easy to install and remove without tools.
  • Supplied as a single reticle for one eyepiece.
Catalog No. NC3833233 Supplier Leica Biosystems Supplier No. 13613561
Reticle designed to install in a compatible eyepiece to provide a 100-division measurement scale for transmitted-light microscopy. Intended for use with Leica 13613532 eyepiece (10x/20 focusing eyepiece), it enables precise linear measurement, particle counting, and consistent reference marking.
  • Provides 100 divisions for precise linear measurement and particle counting.
  • Compatible with focusing eyepieces that accept removable reticles.
  • Improves reproducibility and reference marking during routine microscopy.
  • Easy to install and remove without tools.
  • Supplied as a single reticle for one eyepiece.