Learn More
Bioworld Leifert and Waites Sterility Test Medium, 500 g
Catalog No. 50254638
Encompass_Preferred
Leifert & Waites Sterility Test MediumContains the Micronutrients, Macronutrients, Vitamins, Sucrose, and Plant Growth Regulators required to initiate plant growth and support the growth of any microorganisms in the explantUsed to determine and identify potential pathogens in explantsCytokines and Auxins should be added as needed Plant tissue culture testedWorking Concentration: 38.2 g/LpH: 6.25–7.25Soluble in water
Catalog No. 50-254-638 Supplier Bioworld Supplier No. 306300421
Add to Cart
Add to Cart
Leifert & Waites Sterility Test MediumContains the Micronutrients, Macronutrients, Vitamins, Sucrose, and Plant Growth Regulators required to initiate plant growth and support the growth of any microorganisms in the explantUsed to determine and identify potential pathogens in explantsCytokines and Auxins should be added as needed Plant tissue culture testedWorking Concentration: 38.2 g/LpH: 6.25–7.25Soluble in water