Learn More
Bellco Glass, Inc. Leighton Tube,16x125mm
SDP
Catalog No. 501944678
Encompass_Preferred
Leighton culture tubes are useful for the study of pathogenic effects on viruses on cells as well as sponge matrix culture and nutritional studies. Use cover slips for preparing expectant tissue cultures with leighton culture tubes. MADE IN THE USA White marking area
Catalog No. 50-194-4678 Supplier Bellco Glass, Inc. Supplier No. 190816125
Leighton culture tubes are useful for the study of pathogenic effects on viruses on cells as well as sponge matrix culture and nutritional studies. Use cover slips for preparing expectant tissue cultures with leighton culture tubes. MADE IN THE USA White marking area