Learn More
Bellco Glass, Inc. Leighton Tube,16X150mm
SDP
Catalog No. 501944677
Encompass_Preferred
Leighton culture tubes are useful for the study of pathogenic effects on viruses on cells as well as sponge matrix culture and nutritional studies. Use cover slips for preparing expectant tissue cultures with leighton culture tubes. MADE IN THE USA Tube dimensions 16x150mm
Catalog No. 50-194-4677 Supplier Bellco Glass, Inc. Supplier No. 190616150
Leighton culture tubes are useful for the study of pathogenic effects on viruses on cells as well as sponge matrix culture and nutritional studies. Use cover slips for preparing expectant tissue cultures with leighton culture tubes. MADE IN THE USA Tube dimensions 16x150mm