Learn More
Bellco Glass, Inc. Leighton Tube Rack
SDP
Catalog No. 501944694
Encompass_Preferred
For leighton type tissue tubes. Hinged front of rack locks tubes into position. MADE IN THE USA For tube lengths: 125 to 150mm Tubes per rack: 36
Catalog No. 50-194-4694 Supplier Bellco Glass, Inc. Supplier No. 191700016
For leighton type tissue tubes. Hinged front of rack locks tubes into position. MADE IN THE USA For tube lengths: 125 to 150mm Tubes per rack: 36