Learn More
US Biomax Inc Leiomyosarcoma tissue array, including TNM and clinical stage (AJCC 7th edition), 80 cases/80 cores(core size 1.5mm), replacing SO804
SDP
Catalog No. NC3433580 Shop All US Biomax Inc Products
Encompass
Leiomyosarcoma tissue microarray, containing 80 cases of leiomyosarcoma (24 cases are G1, 47 cases G2, 9 case G3), single core per case
Catalog No. NC3433580 Supplier US Biomax Inc Supplier No. SO804B
Leiomyosarcoma tissue microarray, containing 80 cases of leiomyosarcoma (24 cases are G1, 47 cases G2, 9 case G3), single core per case