Learn More
ATCC Balamuthia mandrillaris; CDC:V039
Catalog No. 502379555
Encompass
This parasitic protozoan was isolated from the brain tissue of a mandrill that died of amebic meningoencephalitis in the San Diego Zoo.
Catalog No. 50-237-9555 Supplier ATCC Supplier No. 50209
This parasitic protozoan was isolated from the brain tissue of a mandrill that died of amebic meningoencephalitis in the San Diego Zoo.