Learn More
Ted Pella Inc Leitsilber 200 Silver Paint, 30g
SDP
Catalog No. NC9368358
Encompass
Leitsilber is fast drying and has a flat surface texture. Silver content 45%, Resistance: 0.02 - 0.04ohms/square. Drying time ~10 minutes at 20°C. Application can be by brush, dipping or spraying. Acetone (#16023) can be used an extender/thinner or as a cleaner to remove the silver paint.
Catalog No. NC9368358 Supplier Ted Pella Inc Supplier No. 16035
Leitsilber is fast drying and has a flat surface texture. Silver content 45%, Resistance: 0.02 - 0.04ohms/square. Drying time ~10 minutes at 20°C. Application can be by brush, dipping or spraying. Acetone (#16023) can be used an extender/thinner or as a cleaner to remove the silver paint.