Learn More
Medchemexpress LLC LEM-14 | 100MG
SDP
Catalog No. 5000229651
Encompass_Preferred
LEM-14 is a potent and selective NSD2 inhibitor with an IC50 of 132 M LEM-14 has very weak activitvagainst NSD1 and has no activity against NSD3 LEM-14 inhibits fibrotic gene expression in ND but not DIO BMDMs LEM-14 combined with ionizing radiation (IR) enhances the apoptosis rate and reduces the colony-formation ability of CRC cells LEM-14 exhibits enhanced anti-tumor efficacy in Balb/c nude mice bearing LoVo cell xenografts when combined with ionizing radiation LEM-14 has the potential for the research of multiple myeloma and colorectal cancer[1][2][3]
Catalog No. 50-002-29651 Supplier Medchemexpress LLC Supplier No. HY114340100MG
LEM-14 is a potent and selective NSD2 inhibitor with an IC50 of 132 M LEM-14 has very weak activitvagainst NSD1 and has no activity against NSD3 LEM-14 inhibits fibrotic gene expression in ND but not DIO BMDMs LEM-14 combined with ionizing radiation (IR) enhances the apoptosis rate and reduces the colony-formation ability of CRC cells LEM-14 exhibits enhanced anti-tumor efficacy in Balb/c nude mice bearing LoVo cell xenografts when combined with ionizing radiation LEM-14 has the potential for the research of multiple myeloma and colorectal cancer[1][2][3]