Learn More
TARGETMOL CHEMICALS INC LEM-14 5MG
SDP
Catalog No. 502579032
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LEM-14 is a potent NSD2-specific inhibitor (IC50132 uM).LEM-14 may be used in the study of multiple myeloma. purity: 98%
Catalog No. 50-257-9032 Supplier TARGETMOL CHEMICALS INC Supplier No. T90065MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. LEM-14 is a potent NSD2-specific inhibitor (IC50132 uM).LEM-14 may be used in the study of multiple myeloma. purity: 98%