Learn More
Electron Microscopy Sciences Lembert Bone Rongeur Straight
SDP
Catalog No. 5030187
Encompass_Preferred
Bone Rongeur. The jaw width is 2.5 mm and the length is 6 1/2". Made from stainless steel with a polished finish.
Catalog No. 50-301-87 Supplier Electron Microscopy Sciences Supplier No. 77982S
Bone Rongeur. The jaw width is 2.5 mm and the length is 6 1/2". Made from stainless steel with a polished finish.