Learn More
Aviva Systems Biology Corp LEMD2 Antibody - middle region (ARP55792_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP55792_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LEMD2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LEMD2. Alternate names: dJ482C21.1, NET25ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-61615