Learn More
Medchemexpress LLC Lemildipine | 94739-29-4 | 99.6% | 457.30 | 50 MG
SDP
Catalog No. 5000400495
Encompass_Preferred
Lemildipine is a new dihydropyridine calcium entry blocker. It exhibits a protective effect on delayed neuronal death (DND) in the hippocampal CA1 subfield after transient forebrain ischemia, with its effect persisting for up to 4 weeks. It also produces a significant decrease in the blood pressure of spontaneously hypertensive rats (SHR).
  • Protective effect on delayed neuronal death (DND)
  • Effect persisted for up to 4 weeks
  • Significant decrease in blood pressure in spontaneously hypertensive rats (SHR)
  • Hypotensive potency 1.4 times stronger than nifedipine
Catalog No. 50-004-00495 Supplier Medchemexpress LLC Supplier No. HY1966350MG
Lemildipine is a new dihydropyridine calcium entry blocker. It exhibits a protective effect on delayed neuronal death (DND) in the hippocampal CA1 subfield after transient forebrain ischemia, with its effect persisting for up to 4 weeks. It also produces a significant decrease in the blood pressure of spontaneously hypertensive rats (SHR).
  • Protective effect on delayed neuronal death (DND)
  • Effect persisted for up to 4 weeks
  • Significant decrease in blood pressure in spontaneously hypertensive rats (SHR)
  • Hypotensive potency 1.4 times stronger than nifedipine