Learn More
MSC Simple Green, Industrial Cleaner Degreaser, Lemon, 1 gal
Catalog No. 502867874
Encompass
This industrial cleaner and degreaser is a concentrated liquid formula designed for heavy-duty cleaning, degreasing, and deodorizing. It effectively cuts through tough grease, oils, and grime on various surfaces and equipment, offering a non-flammable, non-abrasive, and non-corrosive solution.
  • EPA Safer Choice Certified alternative to caustic or acidic cleaners, bleaches, and solvents
  • Removes dirt, grease, oil, and stains from all washable surfaces
  • Can be used manually or with professional cleaning equipment
  • Versatile formula for a wide range of industries
  • Rinses easily without leaving a residue
  • Complies with volatile organic compound laws
  • Water-based formula free of butyl, solvents, and phosphates
  • Mild alkaline pH increases the range of applications and use
Catalog No. 50-286-7874 Supplier MSC Supplier No. 73239071
This industrial cleaner and degreaser is a concentrated liquid formula designed for heavy-duty cleaning, degreasing, and deodorizing. It effectively cuts through tough grease, oils, and grime on various surfaces and equipment, offering a non-flammable, non-abrasive, and non-corrosive solution.
  • EPA Safer Choice Certified alternative to caustic or acidic cleaners, bleaches, and solvents
  • Removes dirt, grease, oil, and stains from all washable surfaces
  • Can be used manually or with professional cleaning equipment
  • Versatile formula for a wide range of industries
  • Rinses easily without leaving a residue
  • Complies with volatile organic compound laws
  • Water-based formula free of butyl, solvents, and phosphates
  • Mild alkaline pH increases the range of applications and use