Learn More
Cole-Parmer Argos Technologies Autoclave Deodorizers, Lemon; 40/Bottle, 6 Bottles/Cs
Catalog No. 501960907 Shop All Cole-Parmer Products
Encompass_Preferred
Absorb and remove autoclave odors, rather than covering them. These sterilizer deodorizers chemically absorb odor-causing molecules, removing unwanted odors from facilities, rather than masking them. Simply place one capsule in an autoclave or incubator with each load. The heat and moisture will then dissolve the capsule and release a clean, fresh scent.
Catalog No. 50-196-0907 Supplier Cole-Parmer Supplier No. 4446828
Absorb and remove autoclave odors, rather than covering them. These sterilizer deodorizers chemically absorb odor-causing molecules, removing unwanted odors from facilities, rather than masking them. Simply place one capsule in an autoclave or incubator with each load. The heat and moisture will then dissolve the capsule and release a clean, fresh scent.