Learn More
Cole-Parmer Argos Technologies Autoclave Deodorizers, Lemon; 40/Bottle
Catalog No. 501960869 Shop All Cole-Parmer Products
Encompass_Preferred
These sterilizer deodorizers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them. Simply place one capsule in your autoclave or incubator with each load. The heat and moisture dissolves the capsule and releases a clean, fresh scent.
Catalog No. 50-196-0869 Supplier Cole-Parmer Supplier No. 2440719
These sterilizer deodorizers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them. Simply place one capsule in your autoclave or incubator with each load. The heat and moisture dissolves the capsule and releases a clean, fresh scent.