Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lemongrass Oil, Spectrum™ Chemical
SDP

Catalog No. 18603778 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
5 kg
Packaging:
Amber Glass Bottle
Steel Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-778 100 mL Amber Glass Bottle
18-603-779 500 mL Amber Glass Bottle
18-603-780 5 kg Steel Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-603-778 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. L1092100ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

L1092, 8007-02-1

Spectrum™ Chemical Lemongrass Oil

Chemical Identifiers

CAS 8007-02-1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mL
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.