Learn More
Stoelting™ Lempert Rongeur
SDP
Catalog No. 10001800 Shop All Stoelting Co Products
Change view
Click to view available options
Model:
Slight curve
Catalog No. Model
10-001-800 Slight curve
1 options
Catalog No. 10-001-800 Supplier Stoelting™ Supplier No. 5216300
Add to Cart
Add to Cart
Stoelting™ Lempert Rongeurs are strongly constructed instruments with a sharp-edged, scoop-shaped tip, used for gouging out bone.
Rongeurs are used to open a window in bone, often in the skull. It is used in neurosurgery and orthopedic surgery to expose areas for operation or to aid in removal of the brain.
Specifications
For Use With (Application) Neuroscience
Length (English) 6.5 in.
Size 2.5 x 25 mm
Model Slight curve