Learn More
Cayman Chemical Lenacapavir, Purity: ≥98%, CAS: 2189684-44-2, 50 mg
SDP
Catalog No. 502942884
Encompass_Preferred
An inhibitor of HIV capsid protein-protein interactions; inhibits viral replication in HIV-1-infected primary human CD4+ T cells and macrophages (mean IC50s = 32 and 56 pM, respectively), two HIV-2 clinical isolates (mean IC50 = 885 pM), and 23 HIV-1 clinical isolates (mean IC50 = 50 pM); prevents stHIV-A19 infection in pigtail macaques at 25 mg/kg, s.c.
Catalog No. 50-294-2884 Supplier Cayman Chemical Supplier No. 4183050 MG
An inhibitor of HIV capsid protein-protein interactions; inhibits viral replication in HIV-1-infected primary human CD4+ T cells and macrophages (mean IC50s = 32 and 56 pM, respectively), two HIV-2 clinical isolates (mean IC50 = 885 pM), and 23 HIV-1 clinical isolates (mean IC50 = 50 pM); prevents stHIV-A19 infection in pigtail macaques at 25 mg/kg, s.c.