Learn More
Medchemexpress LLC HY-111964 5mg Medchemexpress, Lenacapavir CAS:2189684-44-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1119645MG
Encompass_Preferred
Medchemexpress, HY-111964 5mg Lenacapavir CAS:2189684-44-2 Lenacapavir (GS-6207) is a HIV-1 capsid inhibitor. Lenacapavir shows anti-HIV activity with an EC50 of 100 pM in MT-4 cells. Lenacapavir displays a mean EC50 of 50 pM (20-160 pM) against 23 HIV-1 clinical isolates from different subtypes in peripheral blood mononuclear cells (PBMCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8815