Learn More
Cayman Chemical Lenalidomide, 191732-72-6, 100 mg
SDP
Supplier:  Cayman Chemical 14643100
Encompass_Preferred
An analog of thalidomide that, at 10 μM, alters gene expression and cell viability in a range of cancer cell lines; induces proliferation and enhances the functional capacity of T-lymphocytes, amplifying co-stimulatory signaling pathways
Catalog No. NC0600901