Learn More
Medchemexpress LLC Lenalidomide | 191732-72-6 | 99.94% | 25 MG
SDP
Catalog No. 5000359195
Encompass_Preferred
Lenalidomide (CC-5013) is a derivative of Thalidomide that acts as a molecular glue and is an orally active immunomodulator. It functions as a ligand of ubiquitin E3 ligase cereblon (CRBN), leading to the selective ubiquitination and degradation of lymphoid transcription factors IKZF1 and IKZF3 by the CRBN-CRL4 ubiquitin ligase.
  • Inhibits growth of mature B-cell lymphomas, including multiple myeloma.
  • Induces IL-2 release from T cells.
  • Stimulates T cell proliferation, IFN-γ, and IL-2 production.
  • Inhibits pro-inflammatory cytokines (TNF-α, IL-1, IL-6, IL-12) and elevates anti-inflammatory cytokine IL-10 from human PBMCs.
  • Downregulates IL-6 production and augments myeloma cell apoptosis.
  • Functions as a molecular glue between human E3 ubiquitin ligase cereblon and CKIα.
  • Induces ubiquitination and degradation of this kinase, killing leukemic cells by p53 activation.
Catalog No. 50-003-59195 Supplier Medchemexpress LLC Supplier No. 25MGHYA0003R
Lenalidomide (CC-5013) is a derivative of Thalidomide that acts as a molecular glue and is an orally active immunomodulator. It functions as a ligand of ubiquitin E3 ligase cereblon (CRBN), leading to the selective ubiquitination and degradation of lymphoid transcription factors IKZF1 and IKZF3 by the CRBN-CRL4 ubiquitin ligase.
  • Inhibits growth of mature B-cell lymphomas, including multiple myeloma.
  • Induces IL-2 release from T cells.
  • Stimulates T cell proliferation, IFN-γ, and IL-2 production.
  • Inhibits pro-inflammatory cytokines (TNF-α, IL-1, IL-6, IL-12) and elevates anti-inflammatory cytokine IL-10 from human PBMCs.
  • Downregulates IL-6 production and augments myeloma cell apoptosis.
  • Functions as a molecular glue between human E3 ubiquitin ligase cereblon and CKIα.
  • Induces ubiquitination and degradation of this kinase, killing leukemic cells by p53 activation.