Learn More
Medchemexpress LLC Lenalidomide-4-aminomethyl hydrochloride | 444289-05-8 | 90.9% | 309.75 | 100 MG
SDP
Catalog No. 5000352105
Encompass_Preferred
Lenalidomide-4-aminomethyl hydrochloride is a lenalidomide-based cereblon (CRBN) ligand utilized for the recruitment of CRBN protein. It can be linked to a ligand for protein via a linker to form PROTAC.
  • Lenalidomide-based cereblon (CRBN) ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to a ligand for protein by a linker to form PROTAC
Catalog No. 50-003-52105 Supplier Medchemexpress LLC Supplier No. HY138882A100MG
Lenalidomide-4-aminomethyl hydrochloride is a lenalidomide-based cereblon (CRBN) ligand utilized for the recruitment of CRBN protein. It can be linked to a ligand for protein via a linker to form PROTAC.
  • Lenalidomide-based cereblon (CRBN) ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to a ligand for protein by a linker to form PROTAC