Learn More
Medchemexpress LLC Lenalidomide-4-aminomethyl hydrochloride | 444289-05-8 | MFCD32703223 | 90.9% | 309.75 | C14H16ClN3O3 | 5 MG
SDP
Catalog No. 5000222000
Encompass_Preferred
Lenalidomide-4-aminomethyl hydrochloride is a lenalidomide-derived cereblon (CRBN) ligand used in chemical biology research and PROTAC design as an E3 ligase recruiting moiety. It is supplied as a hydrochloride salt solid, molecular weight 309.75, and chemical formula C14H16ClN3O3.
  • Used as a CRBN ligand for targeted protein degradation studies.
  • Suitable for PROTAC synthesis and linker conjugation.
  • Provided as a hydrochloride salt for handling and stability.
  • Characterized by RP-HPLC and LC-MS analytical methods.
  • Offered in small research pack sizes for screening and optimization.
Catalog No. 50-002-22000 Supplier Medchemexpress LLC Supplier No. HY138882A5MG
Lenalidomide-4-aminomethyl hydrochloride is a lenalidomide-derived cereblon (CRBN) ligand used in chemical biology research and PROTAC design as an E3 ligase recruiting moiety. It is supplied as a hydrochloride salt solid, molecular weight 309.75, and chemical formula C14H16ClN3O3.
  • Used as a CRBN ligand for targeted protein degradation studies.
  • Suitable for PROTAC synthesis and linker conjugation.
  • Provided as a hydrochloride salt for handling and stability.
  • Characterized by RP-HPLC and LC-MS analytical methods.
  • Offered in small research pack sizes for screening and optimization.