Learn More
Medchemexpress LLC Lenalidomide 4'-PEG1-amine dihydrochloride | 2624336-86-1 | 98.37% | 419.30 | 5 MG
SDP
Catalog No. 5000395204
Encompass_Preferred
Lenalidomide 4'-PEG1-amine dihydrochloride is a Lenalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be connected to the ligand for protein by a linker to form PROTAC.
  • Lenalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to the ligand for protein by a linker to form PROTAC
  • For research use only
Catalog No. 50-003-95204 Supplier Medchemexpress LLC Supplier No. HY1611285MG
Lenalidomide 4'-PEG1-amine dihydrochloride is a Lenalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be connected to the ligand for protein by a linker to form PROTAC.
  • Lenalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to the ligand for protein by a linker to form PROTAC
  • For research use only