Learn More
Medchemexpress LLC Lenalidomide 4'-PEG1-amine dihydrochloride | 2624336-86-1 | 98.42% | 419.30 | 25 MG
SDP
Catalog No. 5000394319
Encompass_Preferred
Lenalidomide 4'-PEG1-amine dihydrochloride is a Lenalidomide-based Cereblon ligand. It is utilized in the recruitment of the CRBN protein. This compound can be linked to a ligand for protein through a linker to form a PROTAC (Proteolysis-targeting chimera).
  • Lenalidomide-based Cereblon ligand
  • Utilized in the recruitment of the CRBN protein
  • Can be linked to a ligand for protein through a linker to form a PROTAC (Proteolysis-targeting chimera)
  • Intended for research use only
  • Not sold to patients
Catalog No. 50-003-94319 Supplier Medchemexpress LLC Supplier No. 25MGHY161128
Lenalidomide 4'-PEG1-amine dihydrochloride is a Lenalidomide-based Cereblon ligand. It is utilized in the recruitment of the CRBN protein. This compound can be linked to a ligand for protein through a linker to form a PROTAC (Proteolysis-targeting chimera).
  • Lenalidomide-based Cereblon ligand
  • Utilized in the recruitment of the CRBN protein
  • Can be linked to a ligand for protein through a linker to form a PROTAC (Proteolysis-targeting chimera)
  • Intended for research use only
  • Not sold to patients