Learn More
Medchemexpress LLC Lenalidomide 4'-PEG2-amine dihydrochloride | 2624336-88-3 | 99.0% | 463.36 | 25 MG
SDP
Catalog No. 5000394714
Encompass_Preferred
Lenalidomide 4'-PEG2-amine dihydrochloride is a Lenalidomide-based Cereblon ligand that is used in the recruitment of CRBN protein. It can be connected to the ligand for protein by a linker to form PROTAC.
  • Functions as a Lenalidomide-based Cereblon ligand.
  • Used for the recruitment of CRBN protein.
  • Can be linked to a protein ligand to form PROTAC.
Catalog No. 50-003-94714 Supplier Medchemexpress LLC Supplier No. 25MGHY161129
Lenalidomide 4'-PEG2-amine dihydrochloride is a Lenalidomide-based Cereblon ligand that is used in the recruitment of CRBN protein. It can be connected to the ligand for protein by a linker to form PROTAC.
  • Functions as a Lenalidomide-based Cereblon ligand.
  • Used for the recruitment of CRBN protein.
  • Can be linked to a protein ligand to form PROTAC.