Learn More
Medchemexpress LLC Lenalidomide-5-Br | 1010100-26-1 | 99.9% | 323.15 g·mol⁻¹ | C13H11BrN2O3 | 1 G
SDP
Catalog No. 5000210319
Encompass_Preferred
Lenalidomide-5-Br is a lenalidomide-derived cereblon (CRBN) ligand used to recruit the CRBN E3 ubiquitin ligase for chemical biology applications. It is supplied as a research-grade building block for PROTAC design, where it can be conjugated to a protein-targeting ligand via a linker.
  • Lenalidomide-derived CRBN ligand for recruitment of cereblon
  • Suitable for PROTAC design and linker conjugation
  • High purity appropriate for research use
  • Available in multiple pack sizes (1 g-100 g)
  • Molecular formula C13H11BrN2O3, molecular weight 323.15 g·mol⁻¹
Catalog No. 50-002-10319 Supplier Medchemexpress LLC Supplier No. HYW0729541G
Lenalidomide-5-Br is a lenalidomide-derived cereblon (CRBN) ligand used to recruit the CRBN E3 ubiquitin ligase for chemical biology applications. It is supplied as a research-grade building block for PROTAC design, where it can be conjugated to a protein-targeting ligand via a linker.
  • Lenalidomide-derived CRBN ligand for recruitment of cereblon
  • Suitable for PROTAC design and linker conjugation
  • High purity appropriate for research use
  • Available in multiple pack sizes (1 g-100 g)
  • Molecular formula C13H11BrN2O3, molecular weight 323.15 g·mol⁻¹