Learn More
Medchemexpress LLC Lenalidomide-5-Br | 1010100-26-1 | MFCD32263142 | 99.9% | 323.15 g/mol | C13H11BrN2O3 | 10 G
SDP
Catalog No. 5000210241
Encompass_Preferred
Lenalidomide-5-Br is a brominated lenalidomide derivative that acts as a cereblon (CRBN) E3 ligase ligand for use in PROTAC design and chemical biology research.
  • High purity suitable for research use.
  • Supplied as a light blue to blue solid.
  • Available in multiple pack sizes, including 10 g.
  • Stable under recommended storage conditions when stored as powder or in solvent.
  • Provided with a product data sheet and safety data sheet for handling information.
Catalog No. 50-002-10241 Supplier Medchemexpress LLC Supplier No. HYW07295410G
Lenalidomide-5-Br is a brominated lenalidomide derivative that acts as a cereblon (CRBN) E3 ligase ligand for use in PROTAC design and chemical biology research.
  • High purity suitable for research use.
  • Supplied as a light blue to blue solid.
  • Available in multiple pack sizes, including 10 g.
  • Stable under recommended storage conditions when stored as powder or in solvent.
  • Provided with a product data sheet and safety data sheet for handling information.