Learn More
Medchemexpress LLC Lenalidomide 5'-piperazine-4-methylpiperidine hydrochloride | 2520105-60-4 | 98.6% | 498.45 g/mol | C23H31N5O3·2HCl | 10 MG
SDP
Catalog No. 5000288422
Encompass_Preferred
Lenalidomide 5'-piperazine-4-methylpiperidine hydrochloride is a functionalized cereblon (CRBN) ligand supplied as a hydrochloride salt for research use in PROTAC development and related synthetic chemistry. It provides an E3 ligase-binding moiety with a terminal piperidine handle to facilitate conjugation and linker attachment, and is supplied with supporting analytical documentation for research applications.
  • Functionalized CRBN ligand suitable for PROTAC development.
  • Hydrochloride salt provided as a solid for synthetic handling.
  • High reported purity for research applications (≈98.6%).
  • Small-scale research packaging ideal for medicinal chemistry (10 MG).
  • Provides a terminal piperidine handle for conjugation and linker chemistry.
  • Analytical documentation such as COA and SDS available.
Catalog No. 50-002-88422 Supplier Medchemexpress LLC Supplier No. HY15756610MG
Lenalidomide 5'-piperazine-4-methylpiperidine hydrochloride is a functionalized cereblon (CRBN) ligand supplied as a hydrochloride salt for research use in PROTAC development and related synthetic chemistry. It provides an E3 ligase-binding moiety with a terminal piperidine handle to facilitate conjugation and linker attachment, and is supplied with supporting analytical documentation for research applications.
  • Functionalized CRBN ligand suitable for PROTAC development.
  • Hydrochloride salt provided as a solid for synthetic handling.
  • High reported purity for research applications (≈98.6%).
  • Small-scale research packaging ideal for medicinal chemistry (10 MG).
  • Provides a terminal piperidine handle for conjugation and linker chemistry.
  • Analytical documentation such as COA and SDS available.