Learn More
Medchemexpress LLC Lenalidomide-6-f | 2468780-87-0 | 99.63% | 262.24 | 1 ML
SDP
Catalog No. 5000350090
Encompass_Preferred
Lenalidomide-6-F is a cereblon (CRBN) ligand derived from Lenalidomide. It functions in the recruitment of the CRBN protein. This compound can be linked to a target protein ligand via a linker to create a PROTAC (proteolysis-targeting chimera).
  • Ligand for cereblon (CRBN) protein.
  • Used in the formation of PROTACs.
  • PROTACs utilize the ubiquitin-proteasome system for selective protein degradation.
  • For research use only.
  • Available as a solid, white to off-white in appearance.
  • Purity: 99.63%.
Catalog No. 50-003-50090 Supplier Medchemexpress LLC Supplier No. HY13888110MM/1ML
Lenalidomide-6-F is a cereblon (CRBN) ligand derived from Lenalidomide. It functions in the recruitment of the CRBN protein. This compound can be linked to a target protein ligand via a linker to create a PROTAC (proteolysis-targeting chimera).
  • Ligand for cereblon (CRBN) protein.
  • Used in the formation of PROTACs.
  • PROTACs utilize the ubiquitin-proteasome system for selective protein degradation.
  • For research use only.
  • Available as a solid, white to off-white in appearance.
  • Purity: 99.63%.