Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Lenalidomide

Catalog No. 9015585G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mg
1 g
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-155-85G 100 mg
90-155-8100MG 1 g
90-155-81G 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 90-155-85G Supplier Sigma Aldrich Supplier No. 9015585G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Lenalidomide

Specifications

CAS 191732-72-6
Melting Point 265°C to 268 °C
Molecular Formula C13H13N3O3
Linear Formula C13H13N3O3
MDL Number MFCD07772307
Quantity 100 mg
Synonym 1-Oxo-4-amino-2-(2,6-dioxopiperidin-3-yl)isoindole; 3-(4-Amino-1,3-dihydro-1-oxo-2H-isoindol-2-yl)-2,6-piperidinedione; 3-(4-Amino-1-oxoisoindolin-2-yl)piperidine-2,6-dione; E3 Ligase ligand; Ligand for PROTAC(R) research
Molecular Weight (g/mol) 259.26 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.