Learn More
Medchemexpress LLC Lenalidomide-acetylene-C5-COOH | 2429878-72-6 | 99.3% | 100 MG
SDP
Catalog No. 5000340843
Encompass_Preferred
Lenalidomide-acetylene-C5-COOH is a Lenalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be connected to a ligand for protein by a linker to form PROTAC. This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Lenalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to a ligand for protein by a linker to form PROTAC
  • Functions as a click chemistry reagent
  • Contains an alkyne group that undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups
Catalog No. 50-003-40843 Supplier Medchemexpress LLC Supplier No. HY129608100MG
Lenalidomide-acetylene-C5-COOH is a Lenalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be connected to a ligand for protein by a linker to form PROTAC. This compound is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Lenalidomide-based Cereblon ligand
  • Used in the recruitment of CRBN protein
  • Can be connected to a ligand for protein by a linker to form PROTAC
  • Functions as a click chemistry reagent
  • Contains an alkyne group that undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide groups