Learn More
Medchemexpress LLC Lenalidomide-acetylene-C5-COOH | 2429878-72-6 | 99.3% | 25 MG
SDP
Catalog No. 5000340796
Encompass_Preferred
Lenalidomide-acetylene-C5-COOH is a Lenalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be connected to the ligand for protein by a linker to form PROTAC. This product is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is for research use only and not sold to patients.
  • Cereblon ligand-linker conjugate.
  • Recruits CRBN protein.
  • Forms PROTAC when connected to a protein ligand.
  • Click chemistry reagent.
  • Alkyne group enables copper-catalyzed azide-alkyne cycloaddition (CuAAc).
Catalog No. 50-003-40796 Supplier Medchemexpress LLC Supplier No. 25MGHY129608
Lenalidomide-acetylene-C5-COOH is a Lenalidomide-based Cereblon ligand used in the recruitment of CRBN protein. It can be connected to the ligand for protein by a linker to form PROTAC. This product is a click chemistry reagent, containing an Alkyne group that can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups. It is for research use only and not sold to patients.
  • Cereblon ligand-linker conjugate.
  • Recruits CRBN protein.
  • Forms PROTAC when connected to a protein ligand.
  • Click chemistry reagent.
  • Alkyne group enables copper-catalyzed azide-alkyne cycloaddition (CuAAc).