Learn More
Medchemexpress LLC Lenalidomide-acetylene-C5-COOH | 2429878-72-6 | 99.3% | 50 MG
SDP
Catalog No. 5000341296
Encompass_Preferred
Lenalidomide-acetylene-C5-COOH is a Lenalidomide-based Cereblon ligand that recruits CRBN protein. It can be connected to ligands for protein by a linker to form PROTACs. This product also functions as a click chemistry reagent, featuring an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Lenalidomide-based Cereblon ligand
  • Recruits CRBN protein
  • Can be connected to ligands for protein by a linker to form PROTACs
  • Click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules
Catalog No. 50-003-41296 Supplier Medchemexpress LLC Supplier No. HY12960850MG
Lenalidomide-acetylene-C5-COOH is a Lenalidomide-based Cereblon ligand that recruits CRBN protein. It can be connected to ligands for protein by a linker to form PROTACs. This product also functions as a click chemistry reagent, featuring an alkyne group capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules.
  • Lenalidomide-based Cereblon ligand
  • Recruits CRBN protein
  • Can be connected to ligands for protein by a linker to form PROTACs
  • Click chemistry reagent
  • Contains an alkyne group for copper-catalyzed azide-alkyne cycloaddition (CuAAc) with azide-containing molecules