Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lenalidomide-C4-NH2 50mg | 2435715-90-3 | 351.83 g/mol | C17H22ClN3O3 | 50 MG
SDP

Catalog No. 5000230072
Encompass_Preferred

Lenalidomide-C4-NH2 hydrochloride is a lenalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for research use. It is used as a CRBN-recruiting moiety in targeted protein degradation (PROTAC) design and in biochemical and cell-based assays.

  • Serves as a cereblon (CRBN) binding ligand for recruitment of E3 ubiquitin ligase.
  • Provided as the hydrochloride salt for improved handling and solubility.
  • Typical purity ~95.8%, suitable for research applications.
  • Molecular weight 351.83 g/mol and formula C17H22ClN3O3.
  • Soluble in DMSO and compatible with linker conjugation chemistries for PROTAC synthesis.
  • Available in multiple pack sizes for research-scale use.

Catalog No. 50-002-30072 Supplier Medchemexpress LLC Supplier No. HY115446A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lenalidomide-C4-NH2 hydrochloride is a lenalidomide-derived cereblon (CRBN) ligand supplied as the hydrochloride salt for research use. It is used as a CRBN-recruiting moiety in targeted protein degradation (PROTAC) design and in biochemical and cell-based assays.

  • Serves as a cereblon (CRBN) binding ligand for recruitment of E3 ubiquitin ligase.
  • Provided as the hydrochloride salt for improved handling and solubility.
  • Typical purity ~95.8%, suitable for research applications.
  • Molecular weight 351.83 g/mol and formula C17H22ClN3O3.
  • Soluble in DMSO and compatible with linker conjugation chemistries for PROTAC synthesis.
  • Available in multiple pack sizes for research-scale use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.