Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lenalidomide-C4-NH2 5mg | 2435715-90-3 | 5 MG
SDP

Catalog No. 5000307630
Encompass_Preferred

Lenalidomide-C4-NH2 hydrochloride is a lenalidomide-derived cereblon (CRBN) ligand provided as the hydrochloride salt for research use in targeted protein degradation and PROTAC development.

  • Acts as a cereblon ligand for recruitment of CRBN in degrader constructs.
  • Provided as the hydrochloride salt to improve solubility.
  • Reported purity 95.8% suitable for research applications.
  • Molecular weight 351.83 and molecular formula C17H22ClN3O3.
  • Used as a building block in PROTAC and E3 ligase ligand-linker conjugates.
  • Intended for research use only; not for human therapeutic use.

Catalog No. 50-003-07630 Supplier Medchemexpress LLC Supplier No. HY115446A5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lenalidomide-C4-NH2 hydrochloride is a lenalidomide-derived cereblon (CRBN) ligand provided as the hydrochloride salt for research use in targeted protein degradation and PROTAC development.

  • Acts as a cereblon ligand for recruitment of CRBN in degrader constructs.
  • Provided as the hydrochloride salt to improve solubility.
  • Reported purity 95.8% suitable for research applications.
  • Molecular weight 351.83 and molecular formula C17H22ClN3O3.
  • Used as a building block in PROTAC and E3 ligase ligand-linker conjugates.
  • Intended for research use only; not for human therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.