Learn More
Medchemexpress LLC HY-115446A 100mg Medchemexpress, Lenalidomide-C4-NH2 (hydrochloride) CAS: Purity:>98%
SDP
Catalog No. 501932501
Encompass_Preferred
Medchemexpress, HY-115446A 100mg Lenalidomide-C4-NH2 (hydrochloride) CAS: Lenalidomide-C4-NH2 hydrochloride is the Lenalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Lenalidomide-C4-NH2 hydrochloride can be connected to the ligand for protein by a linker to form PROTAC (Compound 24), which has IC 50 s of 0.98 nM and 13.7 nM in inhibition of RS4;11 and MOLM-13 acute leukemia cell lines growth, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2501 Supplier Medchemexpress LLC Supplier No. HY115446A100MG
Add to Cart
Add to Cart
Medchemexpress, HY-115446A 100mg Lenalidomide-C4-NH2 (hydrochloride) CAS: Lenalidomide-C4-NH2 hydrochloride is the Lenalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Lenalidomide-C4-NH2 hydrochloride can be connected to the ligand for protein by a linker to form PROTAC (Compound 24), which has IC 50 s of 0.98 nM and 13.7 nM in inhibition of RS4;11 and MOLM-13 acute leukemia cell lines growth, respectively [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.