Learn More
Medchemexpress LLC Lenalidomide-C5-acid | 2338824-30-7 | 96.36% | 373.40 | 100 MG
SDP
Catalog No. 5000394597
Encompass_Preferred
Lenalidomide-C5-acid is a Lenalidomide-based E3 ligase ligand-linker conjugate that recruits CRBN protein and can be used to synthesize PROTACs. It is designed for high-speed applications and has a range of documented technical specifications.
  • Recruits CRBN protein
  • Can be used to synthesize PROTACs
  • Available in various sizes
Catalog No. 50-003-94597 Supplier Medchemexpress LLC Supplier No. HY160241100MG
Lenalidomide-C5-acid is a Lenalidomide-based E3 ligase ligand-linker conjugate that recruits CRBN protein and can be used to synthesize PROTACs. It is designed for high-speed applications and has a range of documented technical specifications.
  • Recruits CRBN protein
  • Can be used to synthesize PROTACs
  • Available in various sizes