Learn More
Medchemexpress LLC HY-122725B 100mg Medchemexpress, Lenalidomide-C5-NH2 (hydrochloride) CAS:0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY122725B100MG
Encompass_Preferred
Medchemexpress, HY-122725B 100mg Lenalidomide-C5-NH2 (hydrochloride) CAS:0 Lenalidomide-C5-NH2 hydrochloride is the Lenalidomide-based Cereblon ligand used in the recruitment of CRBN protein. Lenalidomide-C5-NH2 can be connected to the ligand for protein by a linker to form PROTACs, such as MDM2 PROTAC degrader[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9933