Learn More
Apexbio Technology LLC Lenalidomide (CC-5013) 191732-72-6 500mg
SDP
Catalog No. 5000061534
Encompass_Preferred
Lenalidomide (CC-5013) a structural analog of thalidomide is a synthetic small-molecule agent exhibiting anti-tumor properties primarily through immune modulation and antiangiogenic activities Mechanistically lenalidomide promotes the activation of immune functions enhances expression of costimulatory molecules on leukemia lymphocytes and restores immunoglobulin synthesis thereby augmenting humoral immune responses Additionally it improves T cell interactions leading to restored immunological synapse formation and function Current research applications involve investigating lenalidomide s therapeutic potential in multiple myeloma myelodysplastic syndrome (MDS) chronic lymphocytic leukemia (CLL) and other lymphoproliferative disorders including non-Hodgkin lymphoma (NHL)
Catalog No. 50-000-61534 Supplier Apexbio Technology LLC Supplier No. A4211500
Lenalidomide (CC-5013) a structural analog of thalidomide is a synthetic small-molecule agent exhibiting anti-tumor properties primarily through immune modulation and antiangiogenic activities Mechanistically lenalidomide promotes the activation of immune functions enhances expression of costimulatory molecules on leukemia lymphocytes and restores immunoglobulin synthesis thereby augmenting humoral immune responses Additionally it improves T cell interactions leading to restored immunological synapse formation and function Current research applications involve investigating lenalidomide s therapeutic potential in multiple myeloma myelodysplastic syndrome (MDS) chronic lymphocytic leukemia (CLL) and other lymphoproliferative disorders including non-Hodgkin lymphoma (NHL)