Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lenalidomide-F | 2359705-88-5 | MFCD31556181 | 99.4% | 262.24 g/mol | C13H11FN2O3 | 50 MG
SDP

Catalog No. 5000301694
Encompass_Preferred

Lenalidomide-F is an E3 ligase ligand (cereblon binder) used in targeted protein degradation research and PROTAC synthesis. It recruits cereblon to enable proteasome-dependent degradation of target proteins and is supplied as a solid research reagent for in vitro and in vivo studies.

  • High purity (reported ~99.4%).
  • Molecular weight: 262.24 g/mol.
  • Chemical formula: C13H11FN2O3.
  • Soluble in DMSO to 5 mg/mL; may require ultrasonic warming to 60°C.
  • Stable at room temperature for years; in solution store at -80°C or -20°C per guidance.
  • Available in multiple pack sizes (25 mg-1 g; larger quantities available on request).

Catalog No. 50-003-01694 Supplier Medchemexpress LLC Supplier No. HYW03923350MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lenalidomide-F is an E3 ligase ligand (cereblon binder) used in targeted protein degradation research and PROTAC synthesis. It recruits cereblon to enable proteasome-dependent degradation of target proteins and is supplied as a solid research reagent for in vitro and in vivo studies.

  • High purity (reported ~99.4%).
  • Molecular weight: 262.24 g/mol.
  • Chemical formula: C13H11FN2O3.
  • Soluble in DMSO to 5 mg/mL; may require ultrasonic warming to 60°C.
  • Stable at room temperature for years; in solution store at -80°C or -20°C per guidance.
  • Available in multiple pack sizes (25 mg-1 g; larger quantities available on request).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.