Learn More
Medchemexpress LLC HY-A0003B 100mg Medchemexpress, Lenalidomide (hemihydrate) CAS:847871-99-2 Purity:>98%
SDP
Catalog No. 502082300
Encompass_Preferred
Medchemexpress, HY-A0003B 100mg Lenalidomide (hemihydrate) CAS:847871-99-2 Lenalidomide hemihydrate (CC-5013 hemihydrate) is a derivative of Thalidomide and an orally active immunomodulator. Lenalidomide hemihydrate (CC-5013 hemihydrate) is a ligand of ubiquitin E3 ligase cereblon (CRBN), and it causes selective ubiquitination and degradation of two lymphoid transcription factors, IKZF1 and IKZF3, by the CRBN-CRL4 ubiquitin ligase. Lenalidomide hemihydrate (CC-5013 hemihydrate) specifically inhibits growth of mature B-cell lymphomas, including multiple myeloma, and induces IL-2 release from T cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2300 Supplier Medchemexpress LLC Supplier No. HYA0003B100MG
Medchemexpress, HY-A0003B 100mg Lenalidomide (hemihydrate) CAS:847871-99-2 Lenalidomide hemihydrate (CC-5013 hemihydrate) is a derivative of Thalidomide and an orally active immunomodulator. Lenalidomide hemihydrate (CC-5013 hemihydrate) is a ligand of ubiquitin E3 ligase cereblon (CRBN), and it causes selective ubiquitination and degradation of two lymphoid transcription factors, IKZF1 and IKZF3, by the CRBN-CRL4 ubiquitin ligase. Lenalidomide hemihydrate (CC-5013 hemihydrate) specifically inhibits growth of mature B-cell lymphomas, including multiple myeloma, and induces IL-2 release from T cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.