Learn More
Medchemexpress LLC HY-130683 5mg Medchemexpress, Lenalidomide-propargyl-C2-NH2 (hydrochloride) CAS: Purity:>98%
SDP
Catalog No. 501969832
Encompass_Preferred
Medchemexpress, HY-130683 5mg Lenalidomide-propargyl-C2-NH2 (HCl) CAS: Lenalidomide-propargyl-C2-NH2 hydrochloride incorporates a cereblon (CRBN) ligand for the E3 ubiquitin ligase, and a linker. Lenalidomide-propargyl-C2-NH2 hydrochloride can be used to design the PROTAC MD-224 (HY-114312). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9832 Supplier Medchemexpress LLC Supplier No. HY1306835MG
Add to Cart
Add to Cart
Medchemexpress, HY-130683 5mg Lenalidomide-propargyl-C2-NH2 (HCl) CAS: Lenalidomide-propargyl-C2-NH2 hydrochloride incorporates a cereblon (CRBN) ligand for the E3 ubiquitin ligase, and a linker. Lenalidomide-propargyl-C2-NH2 hydrochloride can be used to design the PROTAC MD-224 (HY-114312). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.