Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lenalidomide-propargyl-C2-NH2 hydrochloride | 2489242-23-9 | 98.0% | C18H20ClN3O3 | 10MG
SDP

Catalog No. 5000261222
Encompass_Preferred

Lenalidomide-propargyl-C2-NH2 hydrochloride is a lenalidomide-derived cereblon (CRBN) ligand provided as the hydrochloride salt and intended as a building block for targeted protein degradation research and click-chemistry conjugation in PROTAC development.

  • Lenalidomide-derived CRBN ligand for E3 ligase recruitment.
  • Contains a propargyl (alkyne) handle for click chemistry conjugation.
  • Supplied as the hydrochloride salt for improved handling and stability.
  • High purity (98.0%).
  • Chemical formula C18H20ClN3O3; molecular weight 361.82.
  • Suitable for chemical biology and PROTAC design applications.

Catalog No. 50-002-61222 Supplier Medchemexpress LLC Supplier No. HY13068310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lenalidomide-propargyl-C2-NH2 hydrochloride is a lenalidomide-derived cereblon (CRBN) ligand provided as the hydrochloride salt and intended as a building block for targeted protein degradation research and click-chemistry conjugation in PROTAC development.

  • Lenalidomide-derived CRBN ligand for E3 ligase recruitment.
  • Contains a propargyl (alkyne) handle for click chemistry conjugation.
  • Supplied as the hydrochloride salt for improved handling and stability.
  • High purity (98.0%).
  • Chemical formula C18H20ClN3O3; molecular weight 361.82.
  • Suitable for chemical biology and PROTAC design applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.