Learn More
Medchemexpress LLC Lenalidomide-propargyl-C2-NH2 hydrochloride | 2489242-23-9 | 98.01% | 100 MG
SDP
Catalog No. 5000339962
Encompass_Preferred
Lenalidomide-propargyl-C2-NH2 hydrochloride incorporates a cereblon (CRBN) ligand for the E3 ubiquitin ligase, and a linker. It is also a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Incorporates cereblon (CRBN) ligand
  • E3 ubiquitin ligase ligand
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can be used to design PROTAC MD-224
Catalog No. 50-003-39962 Supplier Medchemexpress LLC Supplier No. HY130683100MG
Lenalidomide-propargyl-C2-NH2 hydrochloride incorporates a cereblon (CRBN) ligand for the E3 ubiquitin ligase, and a linker. It is also a click chemistry reagent containing an Alkyne group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing Azide groups.
  • Incorporates cereblon (CRBN) ligand
  • E3 ubiquitin ligase ligand
  • Click chemistry reagent
  • Contains an Alkyne group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Can be used to design PROTAC MD-224